Learn More
DWK Life Sciences Kimble™ Kontes™ Weighing/Transfer Funnels
Product Code. p-3246762
Change view
Click to view available options
Height (Metric):
13 mm
Diameter (Metric)Top:
60 mm
Unit Size:
Each
Product Code. Height (Metric) Diameter (Metric)Top unitSize
15376657 13 mm 60 mm Each
1 options
Product Code. 15376657
Brand: DWK Life Sciences 7476000013
Only null left
Add to basket
Add to basket
For easy transfer of liquids or powders
  • Flat bottom
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 60 mm
Material Borosilicate Glass
Height (Metric) 13 mm
For Use With (Application) Used for easy transfer of liquids or powders
Product Type Transfer/Weighing Funnel