Learn More
DWK Life Sciences Kimble™ Kontes™ PTFE-Plugged Columns
Product Code. 11887453
Change view
Click to view available options
Length (Metric):
150 mm
200 mm
250 mm
300 mm
350 mm
400 mm
500 mm
700 mm
Diameter (Metric) Inner:
11 mm
15 mm
19 mm
22 mm
25 mm
30 mm
41 mm
49 mm
Column Capacity:
10 mL
20 mL
30 mL
40 mL
80 mL
110 mL
130 mL
140 mL
150 mL
190 mL
240 mL
250 mL
300 mL
650 mL
1300 mL
Includes:
2mm Bore PTFE Stopcock Plug
Fritted Disc, 2mm Bore PTFE Stopcock Plug
Unit Size:
Each
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Column Capacity Includes unitSize
11887453 200 mm 15 mm 30 mL 2mm Bore PTFE Stopcock Plug Each
13406349 250 mm 11 mm 20 mL 2mm Bore PTFE Stopcock Plug Each
11857453 150 mm 11 mm 10 mL 2mm Bore PTFE Stopcock Plug Each
11897453 250 mm 15 mm 40 mL 2mm Bore PTFE Stopcock Plug Each
11807463 500 mm 15 mm 80 mL 2mm Bore PTFE Stopcock Plug Each
11807473 500 mm 19 mm 140 mL 2mm Bore PTFE Stopcock Plug Each
11817473 300 mm 22 mm 110 mL 2mm Bore PTFE Stopcock Plug Each
13426349 500 mm 22 mm 190 mL 2mm Bore PTFE Stopcock Plug Each
11897463 300 mm 19 mm 80 mL 2mm Bore PTFE Stopcock Plug Each
11837473 400 mm 30 mm 250 mL 2mm Bore PTFE Stopcock Plug Each
11827473 350 mm 22 mm 130 mL 2mm Bore PTFE Stopcock Plug Each
11847473 500 mm 30 mm 300 mL 2mm Bore PTFE Stopcock Plug Each
13446339 300 mm 11 mm 20 mL Fritted Disc, 2mm Bore PTFE Stopcock Plug Each
13456339 400 mm 19 mm 110 mL Fritted Disc, 2mm Bore PTFE Stopcock Plug Each
11817483 700 mm 49 mm 1300 mL 2mm Bore PTFE Stopcock Plug Each
13436349 500 mm 41 mm 650 mL 2mm Bore PTFE Stopcock Plug Each
13466339 300 mm 22 mm 110 mL Fritted Disc, 2mm Bore PTFE Stopcock Plug Each
13476339 400 mm 22 mm 150 mL Fritted Disc, 2mm Bore PTFE Stopcock Plug Each
11847423 300 mm 25 mm 140 mL Fritted Disc, 2mm Bore PTFE Stopcock Plug Each
13486339 500 mm 25 mm 240 mL Fritted Disc, 2mm Bore PTFE Stopcock Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Product Code. 11887453 Supplier DWK Life Sciences Supplier No. 4205300221
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket
Manufactured from borosilicate glass 3.3
With PTFE stopcock plug
  • Sealed-in, coarse porosity (40 to 60μm) fritted disc supports packing
  • Use glass wool to support packing in columns without fritted disc
  • PTFE plug controls flowrate
  • Packing not included
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Analytical separation
Diameter (Metric) Inner 15 mm
Type Plain Chromatography Column
Column Capacity 30 mL
Length (Metric) 200 mm
Material Glass
Support Material Glass Wool
Includes 2mm Bore PTFE Stopcock Plug