Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- Low to Medium Pressure LC Columns
- DWK Life Sciences KIMBLE™ KONTES™ PTFE-Plugged Columns
DWK Life Sciences KIMBLE™ KONTES™ PTFE-Plugged Columns
Product Code. 11807463 Shop All DWK Life Sciences Products
Change view
Click to view available options
Length (Metric):
150 mm
200 mm
250 mm
300 mm
350 mm
400 mm
500 mm
700 mm
Diameter (Metric) Inner:
11 mm
15 mm
19 mm
22 mm
25 mm
30 mm
41 mm
49 mm
Column Capacity:
10 mL
20 mL
30 mL
40 mL
80 mL
110 mL
130 mL
140 mL
150 mL
190 mL
240 mL
250 mL
300 mL
650 mL
1300 mL
Includes:
2mm Bore PTFE Stopcock Plug
Fritted Disc, 2mm Bore PTFE Stopcock Plug
Packungsgröße:
Each
20 product options available for selection
| Catalog No. | Length (Metric) | Diameter (Metric) Inner | Column Capacity | Includes | Packungsgröße |
|---|---|---|---|---|---|
| 11807463 | 500 mm | 15 mm | 80 mL | 2mm Bore PTFE Stopcock Plug | Each |
| 11887453 | 200 mm | 15 mm | 30 mL | 2mm Bore PTFE Stopcock Plug | Each |
| 13406349 | 250 mm | 11 mm | 20 mL | 2mm Bore PTFE Stopcock Plug | Each |
| 11857453 | 150 mm | 11 mm | 10 mL | 2mm Bore PTFE Stopcock Plug | Each |
| 11897453 | 250 mm | 15 mm | 40 mL | 2mm Bore PTFE Stopcock Plug | Each |
| 11807473 | 500 mm | 19 mm | 140 mL | 2mm Bore PTFE Stopcock Plug | Each |
| 11817473 | 300 mm | 22 mm | 110 mL | 2mm Bore PTFE Stopcock Plug | Each |
| 13426349 | 500 mm | 22 mm | 190 mL | 2mm Bore PTFE Stopcock Plug | Each |
| 11897463 | 300 mm | 19 mm | 80 mL | 2mm Bore PTFE Stopcock Plug | Each |
| 11837473 | 400 mm | 30 mm | 250 mL | 2mm Bore PTFE Stopcock Plug | Each |
| 11827473 | 350 mm | 22 mm | 130 mL | 2mm Bore PTFE Stopcock Plug | Each |
| 11847473 | 500 mm | 30 mm | 300 mL | 2mm Bore PTFE Stopcock Plug | Each |
| 13446339 | 300 mm | 11 mm | 20 mL | Fritted Disc, 2mm Bore PTFE Stopcock Plug | Each |
| 13456339 | 400 mm | 19 mm | 110 mL | Fritted Disc, 2mm Bore PTFE Stopcock Plug | Each |
| 11817483 | 700 mm | 49 mm | 1300 mL | 2mm Bore PTFE Stopcock Plug | Each |
| 13436349 | 500 mm | 41 mm | 650 mL | 2mm Bore PTFE Stopcock Plug | Each |
| 13466339 | 300 mm | 22 mm | 110 mL | Fritted Disc, 2mm Bore PTFE Stopcock Plug | Each |
| 13476339 | 400 mm | 22 mm | 150 mL | Fritted Disc, 2mm Bore PTFE Stopcock Plug | Each |
| 11847423 | 300 mm | 25 mm | 140 mL | Fritted Disc, 2mm Bore PTFE Stopcock Plug | Each |
| 13486339 | 500 mm | 25 mm | 240 mL | Fritted Disc, 2mm Bore PTFE Stopcock Plug | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
This item is not returnable.
View return policy
Product Code.
11807463
Supplier
DWK Life Sciences
Supplier No.
4205300225
-
2220.00 NOK
1709.60 NOK / Pack of 1
Save 510.40 NOK
22% Off
You must Sign In and have a business account to purchase this item.
Restricted Item
Estimated Shipment: 08-06-2027
Log in to see stock.
Log in to see stock.
Add to basket
Add to basket
This item is not returnable.
View return policy
Description
With PTFE stopcock plug
- Sealed-in, coarse porosity (40 to 60μm) fritted disc supports packing
- Use glass wool to support packing in columns without fritted disc
- PTFE plug controls flowrate
- Packing not included
Specifications
Specifications
| Pore Size | 40 to 60 μm |
| Column Capacity | 80 mL |
| Length (Metric) | 500 mm |
| Material | Glass |
| Support Material | Glass Wool |
| Includes | 2mm Bore PTFE Stopcock Plug |
| For Use With (Application) | Analytical separation |
| Type | Plain Chromatography Column |
| Diameter (Metric) Inner | 15 mm |