Learn More
DWK Life Sciences Kimble™ Kontes™ Pellet Pestle™
Product Code. 11849923
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
Material:
CTFE/Stainless Steel
Polypropylene
Includes:
100 Microtubes
w/o Microtubes
Dnase Free:
No
RNase, DNase and Pyrogen-free
Unit Size:
Each
Pack of 100
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Material Includes Dnase Free unitSize
11849923 1.5 mL Polypropylene w/o Microtubes No Pack of 100
11898502 1.5 mL CTFE/Stainless Steel w/o Microtubes No Each
11888502 0.5 mL CTFE/Stainless Steel w/o Microtubes No Each
11872913 1.5 mL Polypropylene 100 Microtubes No Pack of 100
11858502 0.5 mL Polypropylene w/o Microtubes RNase, DNase and Pyrogen-free Pack of 100
10057312 1.5 mL Polypropylene 100 Microtubes RNase, DNase and Pyrogen-free Pack of 100
10794783 0.5 mL Polypropylene 100 Microtubes RNase, DNase and Pyrogen-free Pack of 100
11815125 1.5 mL Polypropylene w/o Microtubes RNase, DNase and Pyrogen-free Pack of 100
11800104 0.5 mL Polypropylene w/o Microtubes No Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11849923 Supplier DWK Life Sciences Supplier No. 7495211500
Trusted Glassware Deals
Use promo code 28509
View all promotions
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
Use to resuspend protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Disposable Pellet Pestle
  • Choose autoclavable polypropylene or RNase-, DNase-, pyrogen-free material (manufactured under M5.5 [Class 10,000] cleanroom conditions)
  • Available with or without 100 microtubes
  • Microtubes have the same properties and come individually wrapped to ensure purity
  • Length of pestle with tip, 7cm (2.75 in.)
Reusable Pellet Pestle
  • Autoclavable CTFE polymeric head with a stainless-steel shaft
  • Pestle length with tip, 6cm (2.5 in.) for 0.5mL size; 14cm (5.59 in.) for 1.5mL size
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color Blue
Capacity (English) 0.05 oz.
Capacity (Metric) 1.5 mL
Material Polypropylene
Product Type Pellet Pestle
Disposable Single-use
For Use With (Application) Resuspend Protein and DNA pellets or grind soft tissue in microcentrifuge tubes
Length (English) 2.75 in.
Dnase Free No
Includes w/o Microtubes
Length (Metric) 70mm
Capacity 0.05 oz., 1.5mL