Learn More
DWK Life Sciences Kimble™ Kontes™ FlexColumn Economy Columns Flow Adapters
Product Code. 11708242
Change view
Click to view available options
For Use With:
10 mm I.D. FlexColumn
15 mm I.D. FlexColumn
25 mm I.D. FlexColumn
Unit Size:
Each
Product Code. For Use With unitSize
11708242 25 mm I.D. FlexColumn Each
11798232 10 mm I.D. FlexColumn Each
13423646 15 mm I.D. FlexColumn Each
3 options
Product Code. 11708242
Brand: DWK Life Sciences 4204152500
Only null left
Add to basket
Add to basket
Convert standard FlexColumn™ columns to adjustable bed volume columns
Includes
  • PTFE eluent cannula
  • Polyacetal body
  • Viton™ O-ring seal
  • One replacement 20μm polyethylene bed support
Order Info
Additional bed supports can be ordered in packs of 10 (see K420415-1001, -1501, -2501).
TRUSTED_SUSTAINABILITY
Specifications
Type Flow Control Adapter
For Use With (Application) Concentration and pH gradients, reverse flow and gravity flow separation
Includes 0.062 in. O.D. FEP PTFE tubing, Viton™ FKM O-ring seal, replacement 20um porosity HDPE bed support
For Use With 25 mm I.D. FlexColumn
Overall Length (Metric) 140 mm
Material Polyacetal, HDPE