Learn More
DWK Life Sciences Kimble™ Kontes™ FlexColumn™ Economy Columns
Product Code. 12746768
Click to view available options
Length (Metric):
40 mm
100 mm
300 mm
500 mm
Column Capacity:
1.5 mL
4 mL
12 mL
19 mL
Quantity:
1
10
Unit Size:
Each
Pack of 10
Product Code. 12746768
Brand: DWK Life Sciences 4204010730
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for use with FPLC™ systems, peristaltic pumps, or gravity flow to 15psi
Use columns with ion-exchange, affinity, gel filtration, and hydrophobic interaction media.
  • Durable borosilicate glass
  • Precision-ground column ends form positive seals with endcaps so they don't leak or “creep” under pressure
  • Threaded, easy to remove reservoir cap won't pop off under pressure
  • Reservoir accepts accessory funnels and glass reservoirs
  • Luer-Lock polypropylene inlet and outlet fittings
  • Bed support has a 20μm polyethylene disc that retains most media for low-pressure aqueous chromatography
  • Columns accept flow adapters for adjusting Flex-Column bed height (order adapters separately)
Flow adapters not available
TRUSTED_SUSTAINABILITY
Specifications
Endcapped Yes
For Use With (Application) Ion exchange, affinity, gel filtration and hydrophobic interaction media
Quantity 1
Temperature (Metric) Range 0 to 50°C
Diameter (Metric) Inner 7 mm
Type Economy Chromatography Column
Max. Pressure 14.5 psi
Pore Size 20 μm
Column Capacity 12 mL
Length (Metric) 300 mm
Material Borosilicate Glass Type I Class A
Support Material HDPE
Connector Types Male luer lock
Cross Sectional Area 0.39cm3
Sterilization Method 2N NaOH or 100% Ethanol