Learn More
DWK Life Sciences Kimble™ Kontes™ Filtration Assemblies: Flasks With Side Arm, No. 2 Hose connection
Borosilicate glass flask with side arm, No. 2 hose connection.
895.00 NOK - 3935.00 NOK
Specifications
Connectors | No. 2 Hose Connection |
---|---|
Description | Filtration Flask |
Material | Borosilicate Glass |
Quantity | 1/Cs. |
Product Code | Brand | Volume (Metric) | Connectors | Standard Taper Joint | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Connectors | Standard Taper Joint | Price | Quantity & Availability | |||
11898492
|
DWK Life Sciences
953710-0000 |
125 mL | No. 2 Hose Connection | 5 |
895.00 NOK
Each |
Estimated Shipment: 01-11-2024
Log in to see stock. |
|||
11818502
|
DWK Life Sciences
953760-0000 |
1000 mL | No. 2 Hose Connection | 8 |
1590.00 NOK
Each |
Estimated Shipment: 01-11-2024
Log in to see stock. |
|||
11883173
|
DWK Life Sciences
953760-4000 |
4000 mL | No. 2 Hose Connection | 8 |
3935.00 NOK
Each |
Estimated Shipment: 22-11-2024
Log in to see stock. |
|||
Description
DWK Life Sciences Kimble Kontes Filtration Assemblies: Flasks With Side Arm, No. 2 Hose connection, Connector Types: No. 2 Hose Connection, Borosilicate Glass, Description: Filtration Flask
Order Info
Does not include rubber stopper and hose
Specifications
No. 2 Hose Connection | |
Filtration Flask | |
Borosilicate Glass | |
1/Cs. |