Learn More
DWK Life Sciences Kimble™ Microfiltration Assembly Hardware
Description
47 mm Glass Support Base
Specifications
Specifications
| For Use With (Equipment) | Ultra-Ware 47mm Microfiltration Assembly |
| Material | Glass |
| Standard Taper Joint | 40/35 |
| Type | Glass Support Base |
| Length (Metric) | 47 mm |