Learn More
DWK Life Sciences Kimble™ Kontes™ Dounce Tissue Grinders
Product Code. 11893133
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1 mL
2 mL
7 mL
15 mL
40 mL
100 mL
Clearance (English):
0.0005 to 0.0025 in.
0.0005 to 0.0055 in.
0.0008 to 0.0022 in.
0.001 to 0.003 in.
Length (English):
1.4 in.
2.3 in.
3.4 in.
4.9 in.
6.1 in.
8.4 in.
9.1 in.
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Clearance (English) Length (English) Unit Size
11893133 100 mL 0.0005 to 0.0055 in. 9.1 in. Each
13495469 2 mL 0.0005 to 0.0025 in. 2.3 in. Each
10145564 15 mL 0.001 to 0.003 in. 6.1 in. Each
10749155 7 mL 0.0008 to 0.0022 in. 4.9 in. Each
11868512 40 mL 0.001 to 0.003 in. 8.4 in. Each
10506645 0.5 mL 0.0005 to 0.0025 in. 1.4 in. Each
13406419 1 mL 0.0005 to 0.0025 in. 3.4 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11893133 Supplier DWK Life Sciences Supplier No. 8853000100
  • ONLINE EXCLUSIVE PRICE
    5430.00 NOK
    4790.22 NOK / Pack of 1
    Save 639.78 NOK
    11% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Designed for cellular work in which the nucleus remains intact after homogenization
All-glass construction
Order Info
Items are sold one per case. Supplied with two pestles: large (for initial sample reduction) and small (to form the final homogenate)
Specifications
Clearance (English) 0.0005 to 0.0055 in.
Clearance (Metric) 0.05 to 0.25mm
Capacity (English) 3.3 oz.
Capacity (Metric) 100 mL
Material Glass
Product Type Dounce Tissue Grinder
For Use With (Application) Designed for cellular work in which the nucleus remains intact after homogenization
Length (English) 9.1 in.
Length (Metric) 232mm