Learn More
DWK Life Sciences Kimble™ Kontes™ Dounce Tissue Grinders
Product Code. 10506645
Change view
Click to view available options
Capacity:
0.01 oz., 0.5mL
0.03 oz., 1mL
0.06 oz., 2mL
0.23 oz., 7mL
0.5 oz., 15mL
1.35 oz., 40mL
3.3 oz., 100mL
Clearance (English):
0.0005 to 0.0025 in.
0.0005 to 0.0055 in.
0.0008 to 0.0022 in.
0.001 to 0.003 in.
Length (English):
1.4 in.
2.3 in.
3.4 in.
4.9 in.
6.1 in.
8.4 in.
9.1 in.
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Clearance (English) Length (English) unitSize
10506645 0.01 oz., 0.5mL 0.0005 to 0.0025 in. 1.4 in. Each
13495469 0.06 oz., 2mL 0.0005 to 0.0025 in. 2.3 in. Each
11893133 3.3 oz., 100mL 0.0005 to 0.0055 in. 9.1 in. Each
10145564 0.5 oz., 15mL 0.001 to 0.003 in. 6.1 in. Each
10749155 0.23 oz., 7mL 0.0008 to 0.0022 in. 4.9 in. Each
11868512 1.35 oz., 40mL 0.001 to 0.003 in. 8.4 in. Each
13406419 0.03 oz., 1mL 0.0005 to 0.0025 in. 3.4 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10506645 Supplier DWK Life Sciences Supplier No. 8853000000
Trusted Glassware Deals
Use promo code 28509
View all promotions
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket
Designed for cellular work in which the nucleus remains intact after homogenization
All-glass construction
Order Info
Items are sold one per case. Supplied with two pestles: large (for initial sample reduction) and small (to form the final homogenate)
TRUSTED_SUSTAINABILITY
Specifications
Clearance (English) 0.0005 to 0.0025 in.
Clearance (Metric) 0.06 to 0.13mm
Capacity (English) 0.01 oz.
Capacity (Metric) 0.5 mL
Material Glass
Product Type Dounce Tissue Grinder
For Use With (Application) Designed for cellular work in which the nucleus remains intact after homogenization
Length (English) 1.4 in.
Length (Metric) 37mm
Capacity 0.01 oz., 0.5mL