Learn More
DWK Life Sciences Kimble™ Kontes™ Dounce Tissue Grinders
Product Code. 10145564
Click to view available options
Capacity:
0.01 oz., 0.5mL
0.03 oz., 1mL
0.06 oz., 2mL
0.23 oz., 7mL
0.5 oz., 15mL
1.35 oz., 40mL
3.3 oz., 100mL
Clearance (English):
0.0005 to 0.0025 in.
0.0005 to 0.0055 in.
0.0008 to 0.0022 in.
0.001 to 0.003 in.
Length (English):
1.4 in.
2.3 in.
3.4 in.
4.9 in.
6.1 in.
8.4 in.
9.1 in.
Unit Size:
Each
Product Code. 10145564
Brand: DWK Life Sciences 8853000015
Only null left
Add to basket
Add to basket
Designed for cellular work in which the nucleus remains intact after homogenization
All-glass construction
Order Info
Items are sold one per case. Supplied with two pestles: large (for initial sample reduction) and small (to form the final homogenate)
TRUSTED_SUSTAINABILITY
Specifications
Clearance (English) 0.001 to 0.003 in.
Clearance (Metric) 0.08 to 0.16mm
Capacity (English) 0.5 oz.
Capacity (Metric) 15 mL
Material Glass
Product Type Dounce Tissue Grinder
For Use With (Application) Designed for cellular work in which the nucleus remains intact after homogenization
Length (English) 6.1 in.
Length (Metric) 157mm
Capacity 0.5 oz., 15mL