Learn More
DWK Life Sciences Kimble™ Kontes™ Replacement Pestle For All-Glass Dounce Tissue Grinders
Product Code. p-3286427
Change view
Click to view available options
Capacity (Metric):
15 mL
Product Type:
Small Clearance Pestle
Unit Size:
Each
Product Code. Capacity (Metric) Product Type unitSize
12058528 15 mL Small Clearance Pestle Each
1 options
Product Code. 12058528
Brand: DWK Life Sciences 8853020015
Only null left
Add to basket
Add to basket
Small and large clearance pestles for 15mL and 40mL Dounce Tissue Grinder.
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 0.5 oz.
Capacity (Metric) 15 mL
Product Type Small Clearance Pestle
For Use With (Application) To form final homogenate
For Use With (Equipment) Dounce all Glass tissue grinders
Capacity 0.5 oz., 15mL