Learn More
DWK Life Sciences Kimble™ Kontes™ Replacement Pestle For All-Glass Dounce Tissue Grinders
Description
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
| Capacity (English) | 0.5 oz. |
| Capacity (Metric) | 15 mL |
| Material | Borosilicate Glass |
| Product Type | Small Clearance Pestle |
| For Use With (Application) | To form final homogenate |
| For Use With (Equipment) | Dounce all Glass tissue grinders |