Learn More
DWK Life Sciences KIMBLE™ KONTES™ Ultra-Ware™ Microfiltration Assembly Component Parts
Product Code. 11893113 Shop All DWK Life Sciences Products
Change view
Click to view available options
Type:
Funnel
Support Base (47 mm)
Support Base (90 mm)
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11893113 Support Base (47 mm) Each
11887523 Support Base (90 mm) Each
11883113 Funnel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11893113 Supplier DWK Life Sciences Supplier No. 9537770001
  • ONLINE EXCLUSIVE PRICE
    2845.00 NOK
    2503.05 NOK / Pack of 1
    Save 341.95 NOK
    12% Off
 Restricted Item
Estimated Shipment: 05-03-2027
KIMBLE™ KONTES™ Ultra-Ware™ Microfiltration Assembly Component Parts are replacement and accessory components designed for use with the Ultra-Ware™ microfiltration assembly system, ensuring reliable filtration performance in laboratory applications.
  • Replacement and accessory components for Kimble™ Kontes™ Ultra-Ware™ microfiltration assemblies
  • Includes glass funnels, support bases, and aluminum clamps to maintain or expand filtration setups
  • Support bases available in 47mm and 90mm sizes to accommodate different filter membrane diameters
  • Aluminum clamp provides secure sealing of funnel to support base during filtration
  • Designed for compatibility with standard Ultra-Ware™ microfiltration assembly configurations
  • Suitable for laboratory filtration of aqueous and compatible solvent samples
Specifications
Disposable Reusable
Material Fritted Glass
Sterility Non-sterile
Type Support Base (47 mm)
Diameter (Metric) 47 mm
For Use With (Equipment) Ultra-ware microfiltration assembly with PTFE-faced fritted glass support