Learn More
DWK Life Sciences Kimble™ KIMAX™ Pycnometer Bottle
Product Code. 12727567
Click to view available options
Capacity (Metric):
25 mL
50 mL
Height (Metric):
175 mm
186 mm
Unit Size:
Each
Product Code. 12727567
Brand: DWK Life Sciences 15123N25
Only null left
Add to basket
Add to basket
Designed for measuring and comparing the densities or specific gravities of liquids and solids. This non-mercury spirit-filled pycnometer is supplied with Standard Taper 10/18 thermometer with a range of 14 to 38°C in 0.2° increments.
  • Thermometer is retested for accuracy within ±0.2 degrees C
  • Standard Taper 5/12 side tube cap has a small vent near the top
  • Body is conical for maximum stability
  • Capacity is within 10 percent of nominal shown on the flask
  • Each flask body is numbered for ready identification
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 25 mL
Height (Metric) 175 mm
Product Type Pycnometer Bottle