Learn More
DWK Life Sciences Kimble™ KIMAX™ Flasks with standard taper Pennyhead Stoppers
Product Code. 12754487
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Size:
13
16
19
22
27
Height (English):
7.08 in.
9.84 in.
11.29 in.
13.3 in.
15.74 in.
Height (Metric):
18 cm
25 cm
28.7 cm
33.8 cm
40 cm
Unit Size:
Pack of 12
Pack of 4
Pack of 6
Product Code. Capacity (Metric) Closure Size Height (English) Height (Metric) unitSize
12754487 1000 mL 22 13.3 in. 33.8 cm Pack of 6
12764487 2000 mL 27 15.74 in. 40 cm Pack of 4
12744487 100 mL 13 7.08 in. 18 cm Pack of 12
12784487 500 mL 19 11.29 in. 28.7 cm Pack of 12
12774487 250 mL 16 9.84 in. 25 cm Pack of 12
5 options
Product Code. 12754487
Brand: DWK Life Sciences 280141000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Calibrated To Contain
Calibrated To Contain
  • Sizes 5mL and larger have designated volumes within Class A tolerances
  • Sizes 1 and 2mL are test-tube shaped
Compliance
Designed from requirements for Class A unserialized flasks of ASTM™ E 288
TRUSTED_SUSTAINABILITY
Specifications
Calibration Calibrated To Contain
Closure Material Glass
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Height (English) 13.3 in.
Height (Metric) 33.8 cm
Tolerance ±0.3 mL
Closure Size 22