Learn More
DWK Life Sciences Kimble™ KIMAX™ Erlenmeyer Flasks with Screw Caps
Product Code. 11717989
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Pack of 2
Pack of 4
Pack of 6
Product Code. Capacity (Metric) unitSize
11717989 2000 mL Pack of 2
11787979 250 mL Pack of 6
11797979 500 mL Pack of 6
11707989 1000 mL Pack of 4
4 options
Product Code. 11717989
Brand: DWK Life Sciences KC267202000
Only null left
Add to basket
Add to basket
Includes
Blue caps and pour rings
Plastic-coated for added safety
  • KimCote™ plastic coating
  • Large openings and GL 45 threaded closures for easy access and tight sealing
  • Blue caps and clear pour rings included
  • Marking spots and scales are durable white ceramic enamel
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Closure Material Polypropylene
Description Reverse metric scale graduations
Diameter (English) Outer 6.34 in.
Capacity (Metric) 2000 mL
Material Borosilicate Glass
Graduation Range 100 to 2000 mL
Height (English) 10.43 in.
Height (Metric) 26.5 cm
Includes Clear pour rings
Writing Surface White Ceramic Enamel Marking
Closure Type Screw
Diameter (Metric) Outer 160 mm