Learn More
DWK Life Sciences Kimble™ KIMAX™ Erlenmeyer Flasks with Screw Caps
Product Code. 11707989
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Pack of 2
Pack of 4
Pack of 6
Product Code. 11707989
Brand: DWK Life Sciences KC267201000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Blue caps and pour rings
Plastic-coated for added safety
  • KimCote™ plastic coating
  • Large openings and GL 45 threaded closures for easy access and tight sealing
  • Blue caps and clear pour rings included
  • Marking spots and scales are durable white ceramic enamel
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Closure Material Polypropylene
Description Reverse metric scale graduations
Diameter (English) Outer 5.04 in.
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Graduation Range 50 to 1000 mL
Height (English) 8.07 in.
Height (Metric) 20.5 cm
Includes Clear pour rings
Writing Surface White Ceramic Enamel Marking
Closure Type Screw
Diameter (Metric) Outer 128 mm