Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Description
- Manufactured from clear 33 expansion borosilicate glass
Specifications
Specifications
| Color | Clear |
| Diameter (Metric) Outer | 19 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 12 mL |
| Thread Style | Screw |
| Volume (English) | 3.2 dr. |
| Closure Size | 15-425 |
| Certifications/Compliance | ASTM Type I, Class A |
| Height (Metric) | 65 mm |