Learn More
DWK Life Sciences KIMBLE™ Clear Screw Thread Sample Vials with Attached PTFE-Faced/White Rubber Lined Caps
Product Code. 11882943
Change view
Click to view available options
Closure Type:
Screw Cap (Closed-top)
Volume (English):
0.5 dr.
1 dr.
10.82 dr.
2.16 dr.
3.2 dr.
4.32 dr.
5.4 dr.
6.5 dr.
6.76 dr.
Unit Size:
Pack of 144
Pack of 288
Pack of 72
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Type Volume (English) Unit Size
11882943 Screw Cap (Closed-top) 10.82 dr. Pack of 72
11813023 Screw Cap (Closed-top) 0.5 dr. Pack of 288
11842963 Screw Cap (Closed-top) 2.16 dr. Pack of 144
11832963 Screw Cap (Closed-top) 1 dr. Pack of 144
11852963 Screw Cap (Closed-top) 3.2 dr. Pack of 144
11862963 Screw Cap (Closed-top) 4.32 dr. Pack of 144
11892943 Screw Cap (Closed-top) 5.4 dr. Pack of 72
11872963 Screw Cap (Closed-top) 6.5 dr. Pack of 144
11802953 Screw Cap (Closed-top) 6.76 dr. Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11882943 Supplier DWK Life Sciences Supplier No. 60811B10
  • ONLINE EXCLUSIVE PRICE
    2656.50 NOK
    2342.43 NOK / Pack of 72
    Save 314.07 NOK
    11% Off
 Restricted Item
Estimated Shipment: 26-01-2027
KIMBLE™ Clear Screw Thread Sample Vials with Attached PTFE-Faced/White Rubber Lined Caps are manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A. Packaged in corrugated trays with partitions.
  • Clear borosilicate glass sample vials for reliable sample storage and handling
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Attached closed-top black phenolic closures with PTFE-faced white rubber liners provide a secure, chemically resistant seal
  • Packaged in corrugated trays with partitions to protect vials during storage and shipping
  • Available in small case quantities for flexible ordering
Specifications
Classification A
Material (Liner) PTFE/Rubber
Color Clear
Diameter (Metric) Outer 28 mm
Material Borosilicate Glass
Pre-slit No
Sterility Non-sterile
Volume (Metric) 40 mL
Thread Style Screw
Volume (English) 10.82 dr.
Closure Material Phenolic
Closure Type Screw Cap (Closed-top)
Quantity 72/Cs.
Marking Spot No
Autoclavable Not Autoclavable
Height (Metric) 95 mm
Product Type Sample Vial with Screw Cap