Learn More
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached PTFE-Faced/White Rubber Lined Caps
Product Code. 11832963
Change view
Click to view available options
Closure Type:
Screw Cap
Screw Cap (Closed-top)
Volume (English):
0.5 dr.
1 dr.
10.82 dr.
2.16 dr.
3.2 dr.
4.32 dr.
5.4 dr.
6.5 dr.
6.76 dr.
Unit Size:
Pack of 144
Pack of 288
Pack of 72
Product Code. Closure Type Volume (English) unitSize
11832963 Screw Cap (Closed-top) 1 dr. Pack of 144
11813023 Screw Cap 0.5 dr. Pack of 288
11842963 Screw Cap (Closed-top) 2.16 dr. Pack of 144
11852963 Screw Cap (Closed-top) 3.2 dr. Pack of 144
11862963 Screw Cap (Closed-top) 4.32 dr. Pack of 144
11892943 Screw Cap (Closed-top) 5.4 dr. Pack of 72
11872963 Screw Cap (Closed-top) 6.5 dr. Pack of 144
11882943 Screw Cap (Closed-top) 10.82 dr. Pack of 72
11802953 Screw Cap (Closed-top) 6.76 dr. Pack of 72
9 options
Product Code. 11832963
Brand: DWK Life Sciences 60811B1
Only null left
Add to basket
Add to basket
Packaged in corrugated trays with partitions
  • Small case quantities
  • Manufactured from clear 33 expansion, borosilicate glass
  • Attached closed top black phenolic closures
TRUSTED_SUSTAINABILITY
Specifications
Classification A
Material (Liner) PTFE/Rubber
Color Clear
Diameter (Metric) Outer 15 mm
Material Borosilicate Glass
Volume (Metric) 4 mL
Thread Style Screw
Volume (English) 1 dr.
Closure Material Phenolic
Closure Type Screw Cap (Closed-top)
Height (Metric) 45 mm