Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Vials w/Attached PTFE-Faced/White Rubber Lined Closures
Product Code. 11833023
Change view
Click to view available options
Volume (Metric):
1.8 mL
2 mL
4 mL
5.5 mL
20 mL
40 mL
Height (Metric):
28 mm
35 mm
38 mm
40 mm
45 mm
57 mm
95 mm
Diameter (Metric) Outer:
12 mm
15 mm
17 mm
19 mm
28 mm
Unit Size:
Pack of 144
Pack of 200
Pack of 288
Pack of 72
Product Code. Volume (Metric) Height (Metric) Diameter (Metric) Outer unitSize
11833023 1.8 mL 35 mm 12 mm Pack of 288
11852973 4 mL 45 mm 15 mm Pack of 144
11833003 2 mL 28 mm 15 mm Pack of 200
11843003 4 mL 38 mm 17 mm Pack of 200
11853003 5.5 mL 40 mm 19 mm Pack of 200
11872953 20 mL 57 mm 28 mm Pack of 72
11862953 40 mL 95 mm 28 mm Pack of 72
7 options
Product Code. 11833023
Brand: DWK Life Sciences 60911B12
Only null left
Add to basket
Add to basket
Attached closed top black phenolic closures
  • Manufactured from amber 51 expansion borosilicate glass
  • Closures with PTFE-faced/white rubber liners or white rubber liners
  • Vials packaged in corrugated trays with partitions
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) PTFE/Rubber
Color Amber
Diameter (Metric) Outer 12 mm
Material Borosilicate Glass
Volume (Metric) 1.8 mL
Thread Style Screw
Volume (English) 0.5 dr.
Closure Material Phenolic
Closure Type Screw Cap (Closed-top)
Certifications/Compliance USP Type I
Height (Metric) 35 mm