Learn More
DWK Life Sciences Kimble™ Amber Plain Disposable Borosilicate Glass Tubes
Product Code. 15826331
Click to view available options
Length (Metric):
100 mm
Diameter (Metric):
16 mm
Unit Size:
Pack of 1000
Product Code. 15826331
Brand: DWK Life Sciences 7350016100A
Only null left
Add to basket
Add to basket
Produced from amber borosilicate glass to give superior protection for light sensitive applications.
  • Amber 51 expansion borosilicate glass offers excellent chemical resistance against sodium leaching, a factor common in soda lime glass products
  • Tubes are packed in convenient trays for ease of use on bench tops and in drawers
  • Trays are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-transit breakage and contamination 
  • No marking spot
  • Designed from ASTM Specification E890
  • Manufactured from amber 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
TRUSTED_SUSTAINABILITY
Specifications
Color Amber
Capacity (Metric) 15 mL
Length (Metric) 100 mm
Material Glass
Disposable Single-use
For Use With Light sensitive applications
Label Type None
Packaging 1000/Cs; Shrink-wrapped trays
Diameter (Metric) 16 mm
Product Type Plain Tube