Learn More
DWK Life Sciences Kimble™ 2mL and 4mL Autosampler Vials without Closures:Screw Threads
Product Code. 11893043
Change view
Click to view available options
Color:
Amber
Clear
Closure Size:
13-425
8-425
Marking Spot:
No
Yes
Product Type:
Autosampler Vial
Unit Size:
Pack of 2000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Closure Size Marking Spot Product Type unitSize
11893043 Clear 8-425 No Autosampler Vial Pack of 2000
11803053 Clear 13-425 No Autosampler Vial Pack of 2000
11813053 Clear 13-425 Yes Autosampler Vial Pack of 2000
11853053 Amber 8-425 No Autosampler Vial Pack of 2000
11863053 Amber 13-425 No Autosampler Vial Pack of 2000
11873053 Amber 13-425 Yes Autosampler Vial Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11893043 Supplier DWK Life Sciences Supplier No. 331232S
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket
Add to basket
Constructed of borosilicate glass
  • Finish: Screw thread
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Diameter (Metric) Outer 12 mm
Material Borosilicate Glass
Volume (Metric) 2.0 mL
For Use With (Application) Chromatography applications
Graduated No
Height (English) 1.25 in.
No. per Pack 100
Closure Type Screw Cap
Quantity 100 Pk.
Closure Size 8-425
Marking Spot No
Height (Metric) 32 mm
Product Type Autosampler Vial