Learn More
DWK Life Sciences Kimble™ 2mL and 4mL Autosampler Vials without Closures:Screw Threads
Product Code. 11853053
Click to view available options
Color:
Amber
Clear
Closure Size:
13-425
8-425
Marking Spot:
No
Yes
Type:
Autosampler Vial
Unit Size:
Pack of 2000
Product Code. 11853053
Brand: DWK Life Sciences 511232SA
Only null left
Add to basket
Add to basket
Constructed of borosilicate glass
  • Finish: Screw thread
TRUSTED_SUSTAINABILITY
Specifications
Color Amber
Diameter (Metric) Outer 12 mm
Material Borosilicate Glass
Volume (Metric) 2.0 mL
For Use With (Application) Chromatography applications
Graduated No
Height (English) 1.25 in.
No. per Pack 100
Closure Type Screw Cap
Height (Metric) 32 mm
Quantity 100 Pk.
Type Autosampler Vial
Closure Size 8-425
Marking Spot No