Learn More
DWK Life Sciences Kimble™ 2mL and 4mL Autosampler Vials without Closures: Crimp Openings
Product Code. 11823053
Click to view available options
Color:
Amber
Clear
Marking Spot:
No
Yes
Neck Style:
Regular
Wide
Unit Size:
Pack of 2000
Product Code. 11823053
Brand: DWK Life Sciences 511232CA
Only null left
Add to basket
Add to basket
Constructed of borosilicate glass
  • O.D. x Length: 12 x 32mm
  • Finish: Crimp
Order Info
100 vials/pack, 20 packs/case. Closures not included.
TRUSTED_SUSTAINABILITY
Specifications
Color Amber
Diameter (Metric) Outer 12 mm
Material Borosilicate Glass
Volume (Metric) 2.0 mL
For Use With (Application) Chromatography applications
Height (English) 1.25 in.
Neck Style Regular
No. per Pack 100
Closure Type Crimp Cap
Height (Metric) 32 mm
Quantity 100/Pk.
Marking Spot No