Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Vials without Closures
Click to view available options
Volume (English):
0.5 dr.
1 dr.
1.5 dr.
10 dr.
2 dr.
5 dr.
Closure Size:
13-425
15-425
24-400
8-425
Unit Size:
Pack of 200
Description
- Amber vials offer protection against harmful light and UV rays for light sensitive products.
Specifications
Specifications
Color | Amber |
Diameter (Metric) Outer | 15 mm |
Material | Borosilicate Glass |
Volume (Metric) | 4 mL |
Thread Style | Screw |
Volume (English) | 1 dr. |
Closure Type | Screw Cap |
Height (Metric) | 45 mm |
Certifications/Compliance | USP Type I |
Closure Size | 13-425 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
DWK Life Sciences Kimble™ AmberScrew Thread Vials without Closures >