Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Vials without Closures
Description
- Amber vials offer protection against harmful light and UV rays for light sensitive products.
Specifications
Specifications
| Color | Amber |
| Diameter (Metric) Outer | 28 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 40 mL |
| Thread Style | Screw |
| Volume (English) | 10 dr. |
| Closure Type | Screw Cap |
| Closure Size | 24-400 |
| Certifications/Compliance | USP Type I |
| Height (Metric) | 95 mm |