Learn More
KleenGuard™ G40 Black Polyurethane Coated Nylon Gloves
Product Code. p-3572423
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
60 pairs
Product Code. Size unitSize
15324252 Small 60 pairs
15334252 Medium 60 pairs
15344252 Large 60 pairs
15354252 X-Large 60 pairs
15364252 2X-Large 60 pairs
5 options
Product Code. 15324252
Brand: KleenGuard™ 13837
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Multi-purpose protective gloves designed for tasks that require high dexterity and fine motor skills. The thin-mil design provides tactile sensitivity, making KleenGuard™ G40 Black Polyurethane Coated Nylon Gloves ideal for performing delicate tasks such as electronics assembly.
  • General purpose protective gloves designed for tasks that require high dexterity and fine motor skills
  • Thin mil design provides tactile sensitivity, ideal for performing delicate tasks
  • Seamless knit nylon backing enhances durability, and provides resistance to tears and abrasion
  • Dark color is ideal for hiding dirt
Specifications
Coated Area Palm, Fingers
Coating Material Polyurethane
Color Black
Exterior Finish Partially Textured
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
Style Full-hand Mitt
For Use With (Application) Small Parts Handling, Precision Assembly, and Inspection
Glove Cuff Style Knit
Series G40
Length (English) 8.7 in.
Length (Metric) 22 cm