Description
- Lightweight “wraparound” design for added comfort and protection
- Impact resistant polycarbonate lens
- Interchangeable head strap allows tight fit to user«s face
- Interchangeable temples can be removed which allows product to be modified to the job
- EN166 N anti-fog coating provides assured protection from fogging lens
- Fog shield provides extra protection around lens
- Cushioned brow bar for additional comfort and ventilation
- Provides 99.9% UV protection, reduces eye strain and protects against radiation
Metal manufacturing, transportation, construction
Compliance
Meets EN166 1FT, CSA and ANSI™ Standards
Specifications
Specifications
| Certifications/Compliance | EN166 1FT, CSA and ANSI Standards |
| Lens Coating | Anti-fog |
| Lens Tint | Clear |
| Product Type | Safety Eyewear |
| Material | Polycarbonate |
| UV Protection | UV400 |