Learn More
DWK Life Sciences Kimble™ KIMAX™ Reusable Tubes with Rubber-Lined Screw Cap
Product Code. 11798263
Change view
Click to view available options
Diameter (Metric):
13 mm
16 mm
20 mm
25 mm
38 mm
Length (Metric):
100 mm
125 mm
150 mm
200 mm
Capacity (Metric):
8 mL
12 mL
16 mL
20 mL
25 mL
30 mL
50 mL
70 mL
160 mL
Thread Size:
13-415
15-415
18-415
24-410
38-430
Unit Size:
Pack of 144
Pack of 192
Pack of 24
Pack of 288
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) Capacity (Metric) Thread Size Unit Size
11798263 20 mm 125 mm 25 mL 18-415 Pack of 192
11728283 25 mm 150 mm 50 mL 24-410 Pack of 144
11738283 25 mm 200 mm 70 mL 24-410 Pack of 144
11768263 16 mm 100 mm 12 mL 15-415 Pack of 288
11708273 20 mm 150 mm 30 mL 18-415 Pack of 192
11748283 38 mm 200 mm 160 mL 38-430 Pack of 24
11778263 16 mm 125 mm 16 mL 15-415 Pack of 288
11788263 16 mm 150 mm 20 mL 15-415 Pack of 288
11758263 13 mm 100 mm 8 mL 13-415 Pack of 288
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11798263 Supplier DWK Life Sciences Supplier No. 4506620125
  • ONLINE EXCLUSIVE PRICE
    6882.75 NOK
    6079.53 NOK / Pack of 192
    Save 803.22 NOK
    11% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Black phenolic cap with inert rubber liner
  • White marking spot
  • Cat. No. 14-930C tube is suitable for determination of insulin according to method of Alving, Rubin, and Millor (Journal of Biological Chemistry [1939], 127, 3)
Specifications
Certifications/Compliance ASTM E982, Type VI, Class A
Closure Color Black
Closure Material Phenolic
Capacity (Metric) 25 mL
Length (Metric) 125 mm
Material Borosilicate Glass
Label Type Marking Spot
Liner Material Rubber
Thread Size 18-415
Diameter (Metric) 20 mm
Closure Type Screw Cap