Learn More
DWK Life Sciences Kimble™ KIMAX™ Precision Bore Burets with PTFE Stopcock
Class A
4545.00 NOK
Specifications
Material | Borosilicate Glass |
---|---|
For Use With (Application) | General Purpose Titrations Requiring Traceable Volumetric Accuracy |
Includes | Dust Cap |
Scale | White |
Stopcock | Straight |
Includes: No. 2 PTFE stopcock and KimKap™ dust cap.
Product Code | Brand | Capacity (Metric) | Graduations | Length (English) | Tolerance | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Graduations | Length (English) | Tolerance | Price | Quantity & Availability | ||
12201368
|
DWK Life Sciences
17027F50 |
50 mL | 0.1 mL | 29.33 in. | ±0.05 mL |
4545.00 NOK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||
Description
Class A
- Precision bore tubing
- Durable white scale
- Permanently marked with individual serial number
- Sold in a Case of 1
Certifications
A certificate is provided to indicate instrument traceability to standards provided by NIST™.
Compliance
- Meet ASTM™ E 287
- 4 1/2 in. (115mm) delivery stem on 10mL size meets ASTM D 664
Specifications
Borosilicate Glass | |
Dust Cap | |
Straight | |
Bore Buret | |
ASTM E287, ASTM E438, ASTM E542, ASTM E694 |
General Purpose Titrations Requiring Traceable Volumetric Accuracy | |
White | |
PTFE | |
Class A |