Learn More
DWK Life Sciences Kimble™ KIMAX™-51 Brand Reusable Serological Pipets with Tooled Opening
Description
Tempered tip with permanent brown scale fused into uniform bore tubing without etchin
Order Info
Plugs not included.
Compliance
Calibrated To Deliver/Blow Out. Designed from ASTM™ E 1044.
Specifications
Specifications
| Certifications/Compliance | ASTM E1044 |
| Material | Borosilicate Glass |
| Graduated | Yes |
| Graduations | 0.01 mL |
| Length (English) | 11.81 in. |
| Tolerance | ±0.008 mL |
| Color | Black |
| Volume (Metric) | 0.2 mL |
| Length (Metric) | 295 mm |
| Type | Serological Pipet |