Learn More
DWK Life Sciences Kimble™ Kim-Kap™ Closures
Product Code. 10626285
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
For Use With (Equipment):
13 mm Plain-end Culture Tubes
16 mm Plain-end Culture Tubes
18 mm Plain-end Culture Tubes
20 mm Plain-end Culture Tubes
25 mm Plain-end Culture Tubes
38 mm Plain-end Culture Tubes
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
Product Code. Color For Use With (Equipment) unitSize
10626285 Green 18 mm Plain-end Culture Tubes Pack of 1000
10799335 Natural 20 mm Plain-end Culture Tubes Pack of 500
11758283 Natural 13 mm Plain-end Culture Tubes Pack of 500
10399924 Natural 16 mm Plain-end Culture Tubes Pack of 1000
10349594 Natural 38 mm Plain-end Culture Tubes Pack of 250
10389354 Red 13 mm Plain-end Culture Tubes Pack of 1000
10616285 Green 13 mm Plain-end Culture Tubes Pack of 1000
10647245 Yellow 13 mm Plain-end Culture Tubes Pack of 1000
10084554 Blue 13 mm Plain-end Culture Tubes Pack of 1000
10024174 Red 16 mm Plain-end Culture Tubes Pack of 1000
10369934 Yellow 16 mm Plain-end Culture Tubes Pack of 1000
10451815 Green 16 mm Plain-end Culture Tubes Pack of 1000
10492375 Blue 18 mm Plain-end Culture Tubes Pack of 1000
10257774 Yellow 18 mm Plain-end Culture Tubes Pack of 1000
10638205 Yellow 25 mm Plain-end Culture Tubes Pack of 500
15 options
Product Code. 10626285
Brand: DWK Life Sciences 7366318
Only null left
Add to basket
Add to basket
Available in natural (clear) or choice of four colors
  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Product Type Push Cap
Autoclavable Autoclavable
Material Polypropylene
Color Green
Sterility Non-sterile
For Use With (Equipment) 18 mm Plain-end Culture Tubes
Disposable Reusable