missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIFC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20790-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody was raised against the C-terminal region of human KIFC1 protein, and recognizes 55-70 kDa of endogenous KIFC1 protein in multiple tissue lysates.
This gene encodes a member of the SWI2/SNF2 family of proteins, which play a critical role in altering protein-DNA interactions. The encoded protein has been shown to have dsDNA-dependent ATPase activity and RNA polymerase II termination activity. This protein interacts with cell division cycle 5-like, associates with human splicing complexes, and plays a role in pre-mRNA splicing.Specifications
| KIFC1 | |
| Polyclonal | |
| Unconjugated | |
| KIFC1 | |
| HSET, KIFC1, kinesin family member C1, Kinesin like protein 2, Kinesin like protein KIFC1, Kinesin related protein HSET, KNSL2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100502766, 294286, 3833 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5XI63, Q9BW19, Q9QWT9 | |
| KIFC1 | |
| KIFC1 Fusion Protein Ag14492 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction