missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF4A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14344-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The function remains unknown.Specifications
KIF4A | |
Polyclonal | |
Unconjugated | |
KIF4A | |
Chromokinesin A, HSA271784, KIF4, KIF4 G1, KIF4A, kinesin family member 4A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16571, 24137, 84393 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95239, P33174 | |
Kif4, Kif4a | |
KIF4A Fusion Protein Ag5682 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |