Learn More
KIF20A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15911-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a glycoprotein expressed on the surface of NK cells, platelets, monocytes and a subset of T cells. It is a member of the Ig-superfamily containing 2 Ig-like domains of the V-set. The protein mediates cellular adhesion of platelets and megakaryocytic cells to vascular endothelial cells. The protein also plays a role in megakaryocytic cell maturation.Specifications
| KIF20A | |
| Polyclonal | |
| Unconjugated | |
| KIF20A | |
| GG10_2, KIF20A, kinesin family member 20A, Kinesin like protein KIF20A, MKLP2, RAB6KIFL, Rabkinesin 6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10112, 19348, 361308 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95235, P97329 | |
| Kif20a | |
| KIF20A Fusion Protein Ag8700 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |