Learn More
KIDINS220 Mouse anti-Human, Clone: 3G2D8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66748-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a transmembrane protein that is preferentially expressed in the nervous system where it controls neuronal cell survival, differentiation into exons and dendrites, and synaptic plasticity. The encoded protein interacts with membrane receptors, cytosolic signaling components, and cytoskeletal proteins, serving as a scaffold that mediates crosstalk between the neurotrophin pathway and several other intracellular signaling pathways. Aberrant expression of this gene is associated with the onset of various neuropsychiatric disorders and neurodegenerative diseases, including Alzheimer′s disease. Naturally occurring mutations in this gene are associated with a syndrome characterized by spastic paraplegia, intellectual disability, nystagmus and obesity. Alternative splicing results in multiple transcript variants.Specifications
| KIDINS220 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9ULH0 | |
| KIDINS220 | |
| KIDINS220 Fusion Protein Ag17624 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 3G2D8 | |
| Unconjugated | |
| KIDINS220 | |
| ARMS, KIAA1250, KIDINS220 | |
| Mouse | |
| Protein A | |
| RUO | |
| 57498 | |
| -20°C | |
| Liquid |