missing translation for 'onlineSavingsMsg'
Learn More

KIAA1468 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18993-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888804

  • 4593.76 NOK / 150µL
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

KIAA1468
Polyclonal
Unconjugated
KIAA1468
HsT3308, HsT885, KIAA1468
Rabbit
Antigen Affinity Chromatography
RUO
227446, 309053, 57614
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q148V7, Q9P260
KIAA1468, Relch
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers