Learn More
KIAA1468 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18993-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
KIAA1468 | |
Polyclonal | |
Unconjugated | |
KIAA1468 | |
HsT3308, HsT885, KIAA1468 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
227446, 309053, 57614 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q148V7, Q9P260 | |
KIAA1468, Relch | |
Peptide | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |