Learn More
KIAA0430 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25233-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a putative peroxisomal protein that appears to be conserved across Euteleostomi. In humans, it may be autoantigenic.Specifications
| KIAA0430 | |
| Polyclonal | |
| Unconjugated | |
| MARF1 | |
| MARF1, A 362G6.1, KIAA0430, Limkain b1, LKAP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 223989, 9665 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BJ34, Q9Y4F3 | |
| MARF1 | |
| MARF1 Fusion Protein Ag18905 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |