missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ki67/MKI67 Antibody [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB110-89717JF525
4607.00 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Ki67/MKI67 Polyclonal antibody specifically detects Ki67/MKI67 in Human,Mouse,Porcine,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Flow (Intracellular),Gene Knock-Out,Immunocytochemistry/ Immunofluorescence
Specifications
| Ki67/MKI67 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 525 | |
| antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
| The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Mouse KI67/MKI67, between aminoacids 1850-1950 (1899-1916) Uniprot# E9PVX6. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
| 4288 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction