Learn More
KHK-IN-1 hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
KHK-IN-1 hydrochloride is a potent ketohexokinase (KHK) inhibitor with IC50 of 12 nM, interacts with Asp-27B in the ATP-binding region of KHK.
Specifications
Specifications
| Chemical Name or Material | KHK-IN-1 hydrochloride |
| Physical Form | Solid |
| CAS | 1303470-48-5 |
| Color | Yellow |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H27ClN8S |
| Solubility Information | DMSO : 25 mg/mL (54.47 mM; Need ultrasonic) ∣H2O : 14.29 mg/mL (31.13 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | [H]Cl.CSC(C=CC=C1)=C1NC2=NC(N3CCNCC3)=NC4=C2N=CN=C4NCC5CC5 |
| Molecular Weight (g/mol) | 459.01 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.