missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KGA/GAM/GAC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23549-1-AP-20UL
This item is not returnable.
View return policy
Description
GLS has 3 isoforms produced by alternative splicing and this antibody is specific to the 3 isoforms (KGA,GAM,GAC) of GLS.
Sahai (1983) demonstrated phosphate-activated glutaminase (EC 3.5.1.2) in human platelets. It is the major enzyme yielding glutamate from glutamine. Significance of the enzyme derives from its possible implication in behavior disturbances in which glutamate acts as a neurotransmitter(Prusiner, 1981). High heritability of platelet glutaminase was indicated by studies of Sahai and Vogel (1983) who found an intraclass correlation coefficient of 0.96 for monozygotic twins and 0.53 for dizygotic twins.Specifications
| KGA/GAM/GAC | |
| Polyclonal | |
| Unconjugated | |
| GLS | |
| AAD20, GAC, GAM, GLS, GLS1, glutaminase, K glutaminase, KGA, KIAA0838, L glutamine amidohydrolase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14660, 24398, 2744 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| D3Z7P3, O94925, P13264 | |
| Gls | |
| KGA/GAM/GAC Fusion Protein Ag20354 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction