Learn More
Thermo Scientific™ KingFisher Apex Accessories
Product Code. 16562442 Shop All Thermo Scientific Products
Click to view available options
Product Type:
24 Combi Head
24 DW Heating Block
96 DW Head
96 DW Heating Block
96 KF Heating Block
96 PCR Head
Combi Head
Heating Block 24
Heating Block 96
Matrix Heating Block
PCR Plate Heating Block
Plate Adapter
Storage Tube Adapter
UV Lamp (Replacement)
Unit Size:
Each
Product Code. 16562442
Brand: Thermo Scientific™ 24079920
KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source.
KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality
  • User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
  • Provides versatile and high-speed purification with reproducible results
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • BindIx PC Software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval
TRUSTED_SUSTAINABILITY
Specifications
Product Type Combi Head
Volume (Metric) Processing KF 96 plate 15 to 200 μL, 96 DW plate 15 to 1000 μL, 96 storage tube 30 to 200 μL
Format 96-well Plate
Capacity 96 Samples
For Use With (Equipment) KingFisher Apex
Magnetic Rods 96
Particle Size (Metric) ca. >1 μm
Type of Magnet Head 96 Combi head
Plate Type 96 KF, 96 DW, 96 storage tube