missing translation for 'onlineSavingsMsg'
Learn More

Ketoconazole, 99.17%, MP Biomedicals™

Product Code. 11421381 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
50mg
Product Code. Quantity unitSize
11421381 1 g 1g
11491371 50 mg 50mg
11401381 100 mg 100mg
11411381 500 mg 500mg
4 options
This item is not returnable. View return policy

Product Code. 11421381

Brand: MP Biomedicals 0215915891

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 65277-42-1
Molecular Formula C26H28Cl2N4O4
Molecular Weight (g/mol) 534.452
MDL Number MFCD00058579
InChI Key XMAYWYJOQHXEEK-SIULDFEJSA-N
Synonym cis-1-Acetyl-4-(4-((2-(2, 4-dichlorophenyl)-2-(1H-1M-idazol-1-ylmethyl)-1, 3-dioxolan-4-yl) methoxy) phenyl)-piperazine, Ketoisdin
PubChem CID 76973198
IUPAC Name 2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl

Specifications

Type Ketoconazole
Physical Form Powder
Color White
Quantity 1 g
Assay Percent Range >99%
Formula Weight 531.44
Chemical Name or Material Ketoconazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.