Learn More
Gibco™ Keratinocyte-SFM, with L-glutamine
GREENER_CHOICE
Product Code. 11580526
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11580526 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11580526 Supplier Gibco™ Supplier No. 17005034
Edge
Keratinocyte-SFM, with L-glutamine
with L-Glutamine
TRUSTED_SUSTAINABILITY
Specifications
Cell Type Epithelial Cells
Form Liquid
Product Type Keratinocyte-SFM Medium
With Additives Glutamine
For Use With (Application) Mammalian cell culture
Quantity 500 mL
Regulatory Status For research use only
Source Human
Content And Storage 2 to 8°C
Description Keratinocyte-SFM Medium, 500mL, Liquid Form, Human Species, With L-Glutamine, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture