missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KEL Mouse anti-Human, Unconjugated, Clone: 1C8B6, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | KEL |
|---|---|
| Clone | 1C8B6 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16859995
|
Proteintech
67393-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16849995
|
Proteintech
67393-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The KEL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KEL. This antibody recognizes human antigen. The KEL antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| KEL | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 3792 | |
| Primary |
| 1C8B6 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human | |
| IgG2a |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title