Learn More
DWK Life Sciences™ KECK™ Tubing Clamp, PBT, KT
Product Code. 10161731 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
Fits tube OD 10 mm
Fits tube OD 14 mm
Fits tube OD 4.5 mm
Fits tube OD 6 mm
Unit Size:
Each
Pack of 100
Product Code. Size unitSize
10161731 Fits tube OD 14 mm Each
15834688 Fits tube OD 4.5 mm Pack of 100
15911581 Fits tube OD 10 mm Pack of 100
15844688 Fits tube OD 6 mm Pack of 100
4 options
Product Code. 10161731
Brand: DWK Life Sciences™ 286338401
Only null left
Add to basket
Add to basket
For infinitely variable regulation of flow rates.
With just one finger, flow rates in hoses can be infinitely adjusted with KECK™ hose clamps. The clamps are available in different colours for the different hose diameters.
  • Continuous regulation of flow rates
  • Handling with one hand possible
  • Color coding of the clamps according to the different hose diameters
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each
Material PBT
For Use With (Equipment) Infinitely Variable Regulation of Flow Rates
For Use With (Application) infinitely variable regulation of flow rates
Size Fits tube OD 14 mm
Color Red