Learn More
DWK Life Sciences KECK™ O-ring
Product Code. p-7189500 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Outside:
6 mm
7.3 mm
Material:
EPDM
VITON
Unit Size:
Each
Pack of 100
Product Code. Diameter (Metric) Outside Material unitSize
11701429 6 mm EPDM Pack of 100
15834698 7.3 mm EPDM Pack of 100
15921591 7.3 mm VITON Pack of 100
10419542 6 mm VITON Each
4 options
Product Code. 11701429
Brand: DWK Life Sciences 286342005
Only null left
Add to basket
Add to basket
Spare part for the KECK™ adapter KA from glass olive to threaded plastic nozzle.
The O-ring is a component and therefore a spare part for the KECK™ Adapter KA from glass olive to plastic olive.
  • The O-ring is a component/spare part for the KECK™ Adapter KA and is fitted between the screw cap and clamping ring for Adapter KA.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Sealing
Quantity 100/Pk.
Material EPDM
Diameter (Metric) Outside 6 mm
Size 3 mm H